MilliporeSigma
  • Recombinant human G protein-coupled lysophosphatidic acid receptors mediate intracellular calcium mobilization.
Recombinant human G protein-coupled lysophosphatidic acid receptors mediate intracellular calcium mobilization.
Molecular pharmacology (1998-11-06)
S An, T Bleu, Y Zheng, E J Goetzl
ABSTRACT
Mobilization of intracellular Ca2+ is a critical cellular response to lysophosphatidic acid (LPA) in many cell types. Recent identification of endothelial differentiation gene (Edg) 2 and Edg4 as subtypes of G protein-coupled receptors for LPA allowed examination of the Ca2+ mobilization mediated specifically by each subtype. To reduce endogenous background levels while enhancing recombinant receptor-specific signals, the aequorin luminescence method was used to quantify cytoplasmic Ca2+ levels. In TAg-Jurkat T cells transiently co-transfected with apoaequorin and human Edg2 or Edg4 cDNA, LPA dose-dependently increased light emission triggered by increased Ca2+ bound to aequorin. N-Palmitoyl-L-serine-phosphoric acid and N-palmitoyl-L-tyrosine-phosphoric acid, which had been previously shown to be antagonists for Xenopus laevis LPA receptors, did not antagonize the Ca2+-mobilizing effects of Edg2 and Edg4. Surprisingly, they acted as agonists or partial agonists for Edg2 and Edg4. The Ca2+ mobilization by Edg2 and Edg4 was further characterized in stable transfectants of rat HTC4 hepatoma cells. By using the fura-2 fluorescence method, a difference in the kinetics of Ca2+ flux with Edg2 and Edg4 was observed. With Edg2, but not Edg4, the initial increase in the Ca2+ concentration was followed by a sustained influx of extracellular Ca2+. The coincident production of inositol phosphates and the inhibition of Ca2+ mobilization by the phospholipase C inhibitor U73122 strongly suggested that Edg2 and Edg4 mobilize Ca2+ through inositol trisphosphate generated by phospholipase C activation. Pertussis toxin almost completely blocked LPA-induced Ca2+ mobilization by Edg2 but only partially blocked that by Edg4, which suggests that Edg2 transduces Ca2+ mobilization largely through pertussis toxin-sensitive Gi proteins, whereas Edg4 requires both Gi and Gq.
Product Description
Avanti
N-P Tyrosine PA, Avanti Research - A Croda Brand 800725P, powder
Avanti
N-P Serine PA, Avanti Research - A Croda Brand 800715P, powder