MilliporeSigma
  • Highly parallel measurements of interaction kinetic constants with a microfabricated optomechanical device.
Highly parallel measurements of interaction kinetic constants with a microfabricated optomechanical device.
Applied physics letters (2009-09-25)
Steven R Bates, Stephen R Quake
ABSTRACT
We used mechanical trapping of molecular interactions to demonstrate a highly parallel approach to measure the kinetics of biomolecular interactions. This approach consumes 25 fmol of material per measurement and permits 320 measurements in a single experiment. We measured association and dissociation curves for the interactions of 6-His and T7 epitope tags with their antibodies, from which we determined the off rates, on rates, and dissociation constants.
Sigma-Aldrich
Anti-c-Myc−Biotin antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture