MilliporeSigma
  • Interaction of DNA with Cationic Lipid Mixtures-Investigation at Langmuir Lipid Monolayers.
Interaction of DNA with Cationic Lipid Mixtures-Investigation at Langmuir Lipid Monolayers.
Langmuir : the ACS journal of surfaces and colloids (2017-09-06)
Christopher Janich, André Hädicke, Udo Bakowsky, Gerald Brezesinski, Christian Wölk
ABSTRACT
Four different binary lipid mixtures composed of a cationic lipid and the zwitterionic colipids DOPE or DPPC, which show different DNA transfer activities in cell culture models, were investigated at the soft air/water interface to identify transfection efficiency determining characteristics. Langmuir films are useful models to investigate the interaction between DNA and lipid mixtures in a two-dimensional model system by using different surface sensitive techniques, namely, epifluorescence microscopy and infrared reflection-absorption spectroscopy. Especially, the effect of adsorbed DNA on the properties of the lipid mixtures has been examined. Distinct differences between the lipid composites were found which are caused by the different colipids of the mixtures. DOPE containing lipid mixtures form fluid monolayers with a uniform distribution of the fluorescent probe in the presence and absence of DNA at physiologically relevant surface pressures. Only at high nonphysiological pressures, the lipid monolayer collapses and phase separation was observed if DNA was present in the subphase. In contrast, DPPC containing lipid mixtures show domains in the liquid condensed phase state in the presence and absence of DNA in the subphase. The adsorption of DNA at the positively charged mixed lipid monolayer induces phase separation which is expressed in the morphology and the point of appearance of these domains.
Product Description
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand