MilliporeSigma
Bimodal Binding of STIL to Plk4 Controls Proper Centriole Copy Number.
Cell reports (2018-06-14)
Midori Ohta, Koki Watanabe, Tomoko Ashikawa, Yuka Nozaki, Satoko Yoshiba, Akatsuki Kimura, Daiju Kitagawa
ABSTRACT
The number of centrioles is tightly controlled to ensure bipolar spindle assembly, which is a prerequisite to maintain genome integrity. However, our understanding of the fundamental principle that governs the formation of a single procentriole per parental centriole is incomplete. Here, we show that the local restriction of Plk4, a master regulator of the procentriole formation, is achieved by a bimodal interaction of STIL with Plk4. We demonstrate that the conserved short coiled-coil region of STIL binds to and protects Plk4 from protein degradation at the site of procentriole formation. On the other hand, the conserved C-terminal region of STIL named truncated in microcephaly (TIM) domain promotes autophosphorylation and degradation of adjacent Plk4 by the direct interaction. Thus, we propose that positive and negative regulation based on the bimodal binding of Plk4 and STIL ensures the formation of a single procentriole per parental centriole.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PLK-4 Antibody, clone 6H5, clone 6H5, from mouse
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)