MilliporeSigma
  • The Effect of Long-Term Administration of Fatty Acid Amide Hydrolase Inhibitor URB597 on Oxidative Metabolism in the Heart of Rats with Primary and Secondary Hypertension.
The Effect of Long-Term Administration of Fatty Acid Amide Hydrolase Inhibitor URB597 on Oxidative Metabolism in the Heart of Rats with Primary and Secondary Hypertension.
Molecules (Basel, Switzerland) (2018-09-19)
Michał Biernacki, Wojciech Łuczaj, Iwona Jarocka-Karpowicz, Ewa Ambrożewicz, Marek Toczek, Elżbieta Skrzydlewska
ABSTRACT
Fatty acid amide hydrolase (FAAH) inhibitor [3-(3-carbamoylphenyl)phenyl] N-cyclohexylcarbamate (URB597) may influence redox balance and blood pressure through the modulation of endocannabinoids levels. Therefore, this study aimed to compare changes in oxidative metabolism and apoptosis in the hearts of rats with spontaneous hypertension (SHR) and secondary hypertension (11-deoxycorticosterone acetate; DOCA-salt rats) treated by URB597 via intraperitoneal injection for 14 days. The results showed that URB597 decreased the activity of NADPH and xanthine oxidases in both groups of rats. Moreover, in the heart of SHR rats, URB597 led to an increase of enzymatic and nonenzymatic antioxidant activity and levels (catalase, vitamin C, glutathione/glutathione disulfide [GSH/GSSG]) and upregulation of the thioredoxin system; however, NRf2 expression was downregulated. The opposite effect in relation to Nrf2 activity and the thioredoxin system was observed in DOCA-salt rats after URB597 administration. Despite improvement in antioxidant parameters, URB597 enhanced oxidative modifications of phospholipids (4-hydroxynonenal and isoprostanes) and proteins (carbonyl groups) in SHR heart, whereas 4-hydroxynonenal and carbonyl groups levels decreased in the heart of DOCA-salt rats. Obtained results suggest that examined lipid mediators are involved in peroxisome proliferator-activated receptors (PPAR)-independent and PPAR-dependent modulation of cardiac inflammatory reactions. Furthermore, decreased expression of pro-apoptotic proteins (Bax and caspase 3 and 9) was observed after URB597 administration in the heart of both groups of hypertensive rats, whereas expression of the antiapoptotic protein (Bcl-2) increased in SHR rats. Long-term administration of URB597 altered cardiac redox status depending on the type of hypertension. URB597 enhanced oxidative metabolism and reduced pro-apoptotic factors in the heart of SHR rats, increasing the probability of heart metabolic disorders occurrence or progression.
Sigma-Aldrich
Benzaldehyde-d6, 98 atom % D
Sigma-Aldrich
Anti-Heme Oxygenase-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PPAR-α antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Bcl-2 antibody produced in mouse, clone Bcl-2-100, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
URB597, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution