MilliporeSigma
  • MKRN2 inhibits migration and invasion of non-small-cell lung cancer by negatively regulating the PI3K/Akt pathway.
MKRN2 inhibits migration and invasion of non-small-cell lung cancer by negatively regulating the PI3K/Akt pathway.
Journal of experimental & clinical cancer research : CR (2018-08-15)
Jun Jiang, Yitong Xu, Hongjiu Ren, Muli Wudu, Qiongzi Wang, Xin Song, Hongbo Su, Xizi Jiang, Lihong Jiang, Xueshan Qiu
ABSTRACT
Makorin RING zinc finger-2 (MKRN2) belongs to the makorin RING zinc finger family and is a novel ubiquitin E3 ligase targeting the p65 subunit of NF-κB to negatively regulate inflammatory responses; however, the relationship between MKRN2 and tumorigenesis remains unclear. In this study, we clarified the role of MKRN2 in non-small cell lung cancer (NSCLC). Tumor specimens collected from 261 NSCLC patients from 2013 to 2017 were retrieved from the Pathology Archive of the First Affiliated Hospital of China Medical University, and we performed assays to evaluate MKRN2 expression and to determine the impact of MKRN2 silencing and overexpression on NSCLC-cell migration and invasion. We demonstrated that MKRN2 expression was associated with lymph node metastasis, p-TNM stage, cancer-cell differentiation, and poor prognosis. By altering the expression of MKRN2 in selected cell lines, we found that MKRN2 inhibited cell migration and invasion through downregulation of the PI3K/Akt pathway. These results suggested that MKRN2 inhibited NSCLC progression by reducing the metastatic potential of cancer cells. Our findings provide critical insight into the association of MKRN2 expression with favorable clinicopathological characteristics in NSCLC patients and suggested that MKRN2 plays a role in inhibiting NSCLC development.
Sigma-Aldrich
Anti-MKRN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution