MilliporeSigma
  • Disterolphospholipids: nonexchangeable lipids and their application to liposomal drug delivery.
Disterolphospholipids: nonexchangeable lipids and their application to liposomal drug delivery.
Angewandte Chemie (International ed. in English) (2009-05-09)
Zhaohua Huang, Mahmoud Reza Jaafari, Francis C Szoka
ABSTRACT
Extreme makeover of cholesterol: Cholesterol exchange is a major reason for the instability of liposomes in blood. The formation of a covalent hybrid between cholesterol and glycerophosphocholine preserves the bilayer-stabilizing effect of free cholesterol but prevents its transfer from the bilayer. Thus, disterolphospholipids (e.g. 1) are valuable new components for liposome formulation.
Product Description
Avanti
DChemsPC, Avanti Research - A Croda Brand 880346P, powder
Avanti
PChemsPC, Avanti Research - A Croda Brand 880343P, powder
Avanti
PChcPC, Avanti Research - A Croda Brand 880345P, powder
Avanti
OChemsPC, Avanti Research - A Croda Brand 880344P, powder