MilliporeSigma
  • Significance of platelet-activating factor acetylhydrolase in patients with non-insulin-dependent (type 2) diabetes mellitus.
Significance of platelet-activating factor acetylhydrolase in patients with non-insulin-dependent (type 2) diabetes mellitus.
Journal of cellular and molecular medicine (2003-03-04)
M Serban, Cristina Tanaseanu, T Kosaka, Cristina Vidulescu, Irina Stoian, Daciana S Marta, S Tanaseanu, Elena Moldoveanu
ABSTRACT
Non-insulin dependent diabetes mellitus (NIDDM) represents an independent risk factor for cardiovascular diseases (CVD), being characterized by a continuous low-grade inflammation and endothelial activation state. Plasma platelet - activating factor - acetylhydrolases (PAF-AHs) are a subgroup of Ca(2+)-independent phospholipase A(2) family (also known as lipoprotein-associated phospholipases A(2)) that hydrolyze and inactivate the lipid mediator platelet-activating factor (PAF) and/or oxidized phospholipids. This enzyme is considered to play an important role in inflammatory diseases and atherosclerosis. The present study aims to investigate the relations between the levels of PAF-AH activity and LDL-cholesterol / HDL-cholesterol (LDL-ch / HDL-ch) ratio in NIDDM patients as compared to controls. serum PAF-AH activity was measured in 50 patients with dyslipidemia, in 50 NIDDM patients and in 50 controls (normal lipid and glucose levels). Total cholesterol, LDL-ch, HDL-ch, triglyceride and blood glucose were determined in all subjects. All NIDDM patients display hiperlipidemia, with increased LDL-ch and triglyceride levels. There is a significant correlation between LDL-ch levels (especially LDL-ch / HDL-ch ratio) and PAF-AH activity in dyslipidemic and NIDDM patients. Diabetic and dyslipidemic patients have an increased plasma PAF-AH activity correlated with their LDL-ch levels and mainly with LDL-ch / HDL-ch ratio. Plasma PAF-AH high levels appear to be important as a risk marker for endothelial dysfunction in patients with NIDDM.
Product Description
Avanti
14:0 NPS PC, 1-myristoyl-2-(4-nitrophenylsuccinyl)-sn-glycero-3-phosphocholine, powder