MilliporeSigma
Lipid droplets go nuclear.
The Journal of cell biology (2016-01-06)
Robert V Farese, Tobias C Walther
ABSTRACT
Lipid droplets (LDs) are sometimes found in the nucleus of some cells. In this issue, Ohsaki et al. (2016. J. Cell Biol. http://dx.doi.org/10.1083/jcb.201507122) show that the nuclear membrane, promyelocytic leukemia bodies, and the protein PML-II play a role in nuclear LD formation, suggesting functional relationships between these structures.
Product Description
Avanti
C4 TopFluor MG, Avanti Research - A Croda Brand 810293P, powder
Avanti
C11 TopFluor MG, Avanti Research - A Croda Brand 810296P, powder
Avanti
18:1-6:0 DNP-C11 TopFluor TG, Avanti Research - A Croda Brand 810272P, powder
Avanti
18:1-C11 TopFluor-C11 TopFluor TG, Avanti Research - A Croda Brand 810271P, powder
Avanti
18:1-18:1-C11 TopFluor TG, Avanti Research - A Croda Brand 810298C
Avanti
18:1-C11 TopFluor DG, Avanti Research - A Croda Brand 810297P, powder