MilliporeSigma
  • Structural study of the response regulator HupR from Rhodobacter capsulatus. Electron microscopy of two-dimensional crystals on a nickel-chelating lipid.
Structural study of the response regulator HupR from Rhodobacter capsulatus. Electron microscopy of two-dimensional crystals on a nickel-chelating lipid.
Journal of molecular biology (1998-02-07)
C Vénien-Bryan, F Balavoine, B Toussaint, C Mioskowski, E A Hewat, B Helme, P M Vignais
ABSTRACT
Two-dimensional crystals of the histidine-tagged-HupR protein, a transcriptional regulator from the photosynthetic bacterium Rhodobacter capsulatus, were obtained upon specific interaction with a Ni2+-chelated lipid monolayer. HupR is a response regulator of the NtrC family; it activates the transcription of the structural genes, hupSLC, of the [NiFe]hydrogenase. The lipid (Ni-NTA-DOGA) uses the metal chelator nitrilotriacetic group as the hydrophilic headgroup and contains unsaturated oleyl tails to provide the fluidity necessary for two-dimensional protein crystallization. A projection map of the full-length protein at 18 A resolution was generated by analysing electron microscopy micrographs of negatively stained crystals. The HupR protein appeared to be dimeric and revealed a characteristic "propeller-like" motif. Each monomer forms an L-shaped structure.
Product Description
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), chloroform
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), powder