MilliporeSigma
  • Mechanotransduction is required for establishing and maintaining mature inner hair cells and regulating efferent innervation.
Mechanotransduction is required for establishing and maintaining mature inner hair cells and regulating efferent innervation.
Nature communications (2018-10-03)
Laura F Corns, Stuart L Johnson, Terri Roberts, Kishani M Ranatunga, Aenea Hendry, Federico Ceriani, Saaid Safieddine, Karen P Steel, Andy Forge, Christine Petit, David N Furness, Corné J Kros, Walter Marcotti
ABSTRACT
In the adult auditory organ, mechanoelectrical transducer (MET) channels are essential for transducing acoustic stimuli into electrical signals. In the absence of incoming sound, a fraction of the MET channels on top of the sensory hair cells are open, resulting in a sustained depolarizing current. By genetically manipulating the in vivo expression of molecular components of the MET apparatus, we show that during pre-hearing stages the MET current is essential for establishing the electrophysiological properties of mature inner hair cells (IHCs). If the MET current is abolished in adult IHCs, they revert into cells showing electrical and morphological features characteristic of pre-hearing IHCs, including the re-establishment of cholinergic efferent innervation. The MET current is thus critical for the maintenance of the functional properties of adult IHCs, implying a degree of plasticity in the mature auditory system in response to the absence of normal transduction of acoustic signals.
Sigma-Aldrich
Linopirdine, ≥98% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG - Atto 488 antibody produced in goat, whole molecule, 1 mg/mL protein