MilliporeSigma
  • ILF2 Is a Regulator of RNA Splicing and DNA Damage Response in 1q21-Amplified Multiple Myeloma.
ILF2 Is a Regulator of RNA Splicing and DNA Damage Response in 1q21-Amplified Multiple Myeloma.
Cancer cell (2017-07-04)
Matteo Marchesini, Yamini Ogoti, Elena Fiorini, Anil Aktas Samur, Luigi Nezi, Marianna D'Anca, Paola Storti, Mehmet Kemal Samur, Irene Ganan-Gomez, Maria Teresa Fulciniti, Nipun Mistry, Shan Jiang, Naran Bao, Valentina Marchica, Antonino Neri, Carlos Bueso-Ramos, Chang-Jiun Wu, Li Zhang, Han Liang, Xinxin Peng, Nicola Giuliani, Giulio Draetta, Karen Clise-Dwyer, Hagop Kantarjian, Nikhil Munshi, Robert Orlowski, Guillermo Garcia-Manero, Ronald A DePinho, Simona Colla
ABSTRACT
Amplification of 1q21 occurs in approximately 30% of de novo and 70% of relapsed multiple myeloma (MM) and is correlated with disease progression and drug resistance. Here, we provide evidence that the 1q21 amplification-driven overexpression of ILF2 in MM promotes tolerance of genomic instability and drives resistance to DNA-damaging agents. Mechanistically, elevated ILF2 expression exerts resistance to genotoxic agents by modulating YB-1 nuclear localization and interaction with the splicing factor U2AF65, which promotes mRNA processing and the stabilization of transcripts involved in homologous recombination in response to DNA damage. The intimate link between 1q21-amplified ILF2 and the regulation of RNA splicing of DNA repair genes may be exploited to optimize the use of DNA-damaging agents in patients with high-risk MM.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-RALY Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.