MilliporeSigma
  • Efficient cytoplasmic delivery of a fluorescent dye by pH-sensitive immunoliposomes.
Efficient cytoplasmic delivery of a fluorescent dye by pH-sensitive immunoliposomes.
The Journal of cell biology (1985-08-01)
J Connor, L Huang
ABSTRACT
We previously showed that liposomes composed of dioleoylphosphatidyl-ethanolamine and palmitoyl-homocysteine (8:2) are highly fusion competent when exposed to an acidic environment of pH less than 6.5. (Connor, J., M. B. Yatvin, and L. Huang, 1984, Proc. Natl. Acad. Sci. USA. 81:1715-1718). Palmitoyl anti-H2Kk was incorporated into these pH-sensitive liposomes by a modified reserve-phase evaporation method. Mouse L929 cells (k haplotype) treated with immunoliposomes composed of dioleoylphosphatidylethanolamine/palmitoyl-homocysteine (8:2) with an entrapped fluorescent dye, calcein, showed diffused fluorescence throughout the cytoplasm. Measurements by use of a microscope-associated photometer gave an approximate value of 50 microM for the cytoplasmic calcein concentration. This concentration represents an efficient delivery of the aqueous content of the immunoliposome. Cells treated with immunoliposomes composed of dioleoylphosphatidylcholine (pH-insensitive liposomes) showed only punctate fluorescence. The cytoplasmic delivery of calcein by the pH-sensitive immunoliposomes could be inhibited by chloroquine or by incubation at 20 degrees C. These results suggest that the efficient cytoplasmic delivery involves the endocytic pathway, particularly the acidic organelles such as the endosomes and/or lysosomes. One possibility is that the immunoliposomes fuse with the endosome membranes from within the endosomes, thus releasing the contents into the cytoplasm. This nontoxic method should be widely applicable to the intracellular delivery of biomolecules into living cells.
Product Description
Avanti
N-palmitoyl homocysteine (PHC), Avanti Research - A Croda Brand 870320P, powder