MilliporeSigma
  • Effects of taxol resistance gene 1 on the cisplatin response in gastric cancer.
Effects of taxol resistance gene 1 on the cisplatin response in gastric cancer.
Oncology letters (2018-05-29)
Shuquan Duan, Jie Yin, Zhigang Bai, Zhongtao Zhang
ABSTRACT
Cisplatin is used to treat multiple types of solid tumor, including gastric cancer. Although cisplatin initially exhibits good efficacy, therapeutic failure often occurs owing to the development of chemoresistance. To the best of our knowledge, the underlying mechanism of cisplatin resistance remains unknown. The aim of the present study was to assess whether taxol resistance gene 1 (TXR1) has a role in cisplatin response in gastric cancer. The expression of TXR1 in fresh-frozen tissues of patients with gastric cancer who were sensitive or resistance to cisplatin was assessed. The level of TXR1 expression was significantly higher in cisplatin-resistant specimens than that in cisplatin-sensitive specimens. Next, the gastric cancer SGC-7901 cell line was exposed to cisplatin to establish a cisplatin-resistance subline, termed SGC-7901/DDP, which exhibited a 6-fold increases in the level of resistance. TXR1 expression was elevated in SGC-7901/DDP cells. Overexpression of TXR1 induced cisplatin resistance in SGC-7901 cells. Downregulation of TXR1 reversed the drug resistance caused by elevation of TXR1 expression in SGC-7901/DDP cells. Animal experiments proved the effect of TXR1 in inducing cisplatin resistance in vivo. Further investigation revealed that TXR1 regulated cisplatin resistance via apoptosis. In conclusion, TXR1 is worthy of further in-depth study as a potential therapeutic target in patients with gastric cancer.
Sigma-Aldrich
Anti-PRR13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution