MilliporeSigma
  • [1,3]-Transfer of chirality during the Nicholas reaction in gamma-benzyloxy propargylic alcohols.
[1,3]-Transfer of chirality during the Nicholas reaction in gamma-benzyloxy propargylic alcohols.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-01-10)
David D Díaz, Miguel A Ramírez, Víctor S Martín
ABSTRACT
A highly regio- and stereoselective intramolecular [1,5]-hydrogen-transfer process is described. Treatment of gamma-benzyl-protected Co2(CO)6-alpha,gamma-acetylenic diols with BF3 x OEt2 provides bis-homopropargylic alcohols. The reaction occurs within seconds, tolerates a wide range of functionalities, and provides good yields. When the ether group is located at a stereochemically defined carbon atom, the rearrangement occurs with high stereoselectivity, transferring the chirality of the carbinol center to the newly created stereocenter. The cleavage of the benzyloxy group is totally regioselective when additional benzyl ethers are present. The scope and limitations of this novel process in densely substituted substrates are evaluated, and possible competitive reactions and/or stereochemical influences are also described. A mechanism based on a highly ordered chair-like transition state substantiated by a theoretical study is also included.