MilliporeSigma
  • A metal-organic framework with entatic metal centers exhibiting high gas adsorption affinity.
A metal-organic framework with entatic metal centers exhibiting high gas adsorption affinity.
Journal of the American Chemical Society (2006-09-07)
Shengqian Ma, Hong-Cai Zhou
ABSTRACT
A metal-organic framework, PCN-9, containing entatic metal centers, has been synthesized and crystallographically characterized. The H2 and CH4 adsorption enthalpies of PCN-9 are among the highest reported thus far.
Sigma-Aldrich
4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid, 95%