MilliporeSigma
  • Oncogenic IL7R is downregulated by histone deacetylase inhibitor in esophageal squamous cell carcinoma via modulation of acetylated FOXO1.
Oncogenic IL7R is downregulated by histone deacetylase inhibitor in esophageal squamous cell carcinoma via modulation of acetylated FOXO1.
International journal of oncology (2018-05-12)
Myoung Jun Kim, Sung Kyung Choi, Seong Hwi Hong, Jung Woo Eun, Suk Woo Nam, Jeung-Whan Han, Jueng Soo You
ABSTRACT
The interleukin-7 receptor (IL7R) is generally expressed in immune cells and is critical in survival, development and homeostasis in the immune system. Advanced genome-wide cancer studies have reported that IL7R is genetically amplified in human esophageal squamous cell carcinoma (ESCC), however, the exact role of IL7R in ESCC has not been investigated. In the present study, it was found that IL7R was overexpressed in ESCC cohorts and the loss of IL7R induced anti-oncogenic effects in ESCC cell lines. A small panel of epigenetic drugs were screened for their ability to downregulate the expression of IL7R. Unexpectedly, apicidin, a histone deacetylase (HDAC) inhibitor, effectively downregulated the expression of IL7R in a dose-dependent manner at an early time-point, as determined by quantitative polymerase chain reaction and IL7R immunostaining, and did not require de novo protein synthesis. Of note, apicidin induced the acetylation of Forkhead box-containing protein, O subfamily 1, which acts as a repressor at the IL7R promoter, accompanied with depleted active histone modifications based on chromatin immunoprecipitation assay. Taken together, these results demonstrated that targeting oncogenic IL7R in ESCC by HDAC inhibitors may be a valuable therapeutic approach.
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human IL7R
Sigma-Aldrich
BIX, ≥98% (HPLC)
Sigma-Aldrich
Histone Acetyltransferase p300 Inhibitor, C646, Histone Acetyltransferase p300 Inhibitor, C646, CAS 328968-36-1, is a cell-permeable, reversible inhibitor of p300/CBP HAT (Ki = 400 nM). Competes with acetyl-CoA for the p300 Lys-CoA binding pocket.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit