MilliporeSigma
  • S-protected thiolated cyclodextrins as mucoadhesive oligomers for drug delivery.
S-protected thiolated cyclodextrins as mucoadhesive oligomers for drug delivery.
Journal of colloid and interface science (2018-07-24)
Mulazim Hussain Asim, Ali Moghadam, Muhammad Ijaz, Arshad Mahmood, Roman Xaver Götz, Barbara Matuszczak, Andreas Bernkop-Schnürch
ABSTRACT
The purpose of this study was to develop a novel mucoadhesive thiolated and S-protected gamma cyclodextrin (γ-CD) with an intact ring backbone to assure a prolonged residence time at specific target sites. Thiolated γ-CD was generated through bromine substitution of its hydroxyl groups followed by replacement to thiol groups using thiourea. In the second step, thiol groups were protected by disulfide bond formation with 2-mercaptonicotinic acid (2-MNA). Thiolated γ-CD displayed 1385 ± 84 µmol thiol groups per gram of oligomer and the amount of MNA determined in the S-protected oligomer was 1153 ± 41 µmol per gram of oligomer. In-vitro screening of mucoadhesive properties of thiolated and S-protected γ-CD was done by two methods. Rheological investigation revealed the conjugates non-mucolytic with only a slight increase in viscosity of thiolated and S-protected γ-CD as compared to unmodified γ-CD, whereas mucoadhesive properties of the new thiolated and S-protected γ-CD performed on freshly excised porcine intestinal mucosa showed 44.4- and 50.9-fold improvement in mucoadhesion, respectively. The new conjugates did not show any cytotoxicity to Caco-2 cells even at a concentration of 1% (m/v) for 24 h. In addition, in-vitro studies of α-amylase degradation of γ-CD, γ-CD-SH and γ-CD-SS-MNA confirmed that all conjugates are biodegradable. These outcomes predict that these new conjugates of γ-CD might provide a new favorable tool for drug delivery providing a prolonged residence time on mucosal surfaces.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%