MilliporeSigma
  • Paeonol-Loaded Ethosomes as Transdermal Delivery Carriers: Design, Preparation and Evaluation.
Paeonol-Loaded Ethosomes as Transdermal Delivery Carriers: Design, Preparation and Evaluation.
Molecules (Basel, Switzerland) (2018-07-19)
Hongdan Ma, Dongyan Guo, Yu Fan, Jing Wang, Jiangxue Cheng, Xiaofei Zhang
ABSTRACT
Paeonol exhibits a wide range of pharmacological activities, such as anti-inflammatory, antidiabetic as well as pain-relieving activities. However, its intrinsic properties, such as low water solubility, poor stability and low oral bioavailability, restrict its clinical application. The current study aimed to optimize paeonol-loaded ethosomal formulation and characterize it in terms of encapsulation efficiency (EE), vesicle size (VS), zeta potential (ZP) and polydispersity index (PDI), in addition to differential scanning calorimetry (DSC), X-ray diffraction (XRD) and Fourier-transform infrared spectroscopy (FT-IR) studies. Here, paeonol-loaded ethosomes were prepared by the injection method and optimized by the single-factor test and central composite design-response surface methodology. The optimized paeonol-loaded ethosomes had an EE of 84.33 ± 1.34%, VS of 120.2 ± 1.3 nm, negative charge of -16.8 ± 0.36 mV, and PDI of 0.131 ± 0.006. Ethosomes showed a spherical morphology under the transmission electron microscope (TEM). DSC, XRD and FT-IR results indicated that paeonol was successfully incorporated into the ethosomes. In-vitro transdermal absorption and skin retention of paeonol from paeonol-loaded ethosomes were 138.58 ± 9.60 µg/cm² and 52.60 ± 7.90 µg/cm², respectively. With reasonable skin tolerance, ethosomes could be a promising vehicle for transdermal delivery of paeonol.
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass