MilliporeSigma
  • Mass Spectrometric Imaging of GABA in the Drosophila melanogaster Adult Head.
Mass Spectrometric Imaging of GABA in the Drosophila melanogaster Adult Head.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2018-07-31)
Yosuke Enomoto, Phan Nt An, Masamitsu Yamaguchi, Eiichiro Fukusaki, Shuichi Shimma
ABSTRACT
Drosophila melanogaster is a model organism in neurodegenerative disease research. In neurodegenerative study, the direct spatial information of neurotransmitters such as γ-aminobutyric acid (GABA) in the brain of Drosophila melanogaster is important to understand the role of GABA. Matrix-assisted laser desorption/ionization imaging mass spectrometry (MALDI-IMS) is an attractive method for direct visualization of neurotransmitters. In this paper, we describe methods to visualize GABA in the brain and head of Drosophila melanogaster using MALDI-IMS.
Product Description
Supelco
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard