MilliporeSigma
  • Fluorescence chemical sensor for determining trace levels of nitroaromatic explosives in water based on conjugated polymer with guanidinium side groups.
Fluorescence chemical sensor for determining trace levels of nitroaromatic explosives in water based on conjugated polymer with guanidinium side groups.
Talanta (2018-06-02)
Hong-Yu Mi, Ji-Lin Liu, Ming-Ming Guan, Qing-Wen Liu, Zhi-Quan Zhang, Guo-Dong Feng
ABSTRACT
A novel fluorescent conjugated polymer (poly(2-amino-N-(2-((4-ethynylphenyl) ethynyl) phenyl)-5-guanidinopentanamide)-1,4-phenylethynylene-1,4-phenyleneethynylene, PPE-Arg) was synthesized in this paper. We found that PPE-Arg could be quenched by picric acid (PA). Photoinduced electron transfer (PET) mechanism can be used to describe the fluorescence quenching of PPE-Arg. It could be speculated that the photo-induced electrons may be transferred from PPE-Arg to nitroaromatic explosives. In this paper, the experiment conditions and detection performance of PPE-Arg were systematically studied. The experiment results demonstrate PPE-Arg as a sensor for PA has a good linear range from 5 × 10-7 to 6 × 10-5 mol L-1 with the calculated limit of detection (LOD) to be 1.0 × 10-7 mol L-1. Meanwhile, reaction time between PPE-Arg and PA is less than 1 min. This proposed sensor was applied to rapidly detect nitroaromatic explosives in environmental water samples and satisfactory results were obtained.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%