MilliporeSigma
  • Synthesis and biological activity of a CXCR4-targeting bis(cyclam) lipid.
Synthesis and biological activity of a CXCR4-targeting bis(cyclam) lipid.
Organic & biomolecular chemistry (2018-08-30)
Anna D Peters, Catriona McCallion, Andrew Booth, Julie A Adams, Karen Rees-Unwin, Alain Pluen, John Burthem, Simon J Webb
ABSTRACT
A bis(cyclam)-capped cholesterol lipid designed to bind C-X-C chemokine receptor type 4 (CXCR4) was synthesised in good overall yield from 4-methoxyphenol through a seven step synthetic route, which also provided a bis(cyclam) intermediate bearing an octaethyleneglycol-primary amine that can be easily derivatised. This bis(cyclam)-capped cholesterol lipid was water soluble and self-assembled into micellar and non-micellar aggregates in water at concentrations above 8 μM. The bioactivity of the bis(cyclam)-capped cholesterol lipid was assessed using primary chronic lymphocytic leukaemia (CLL) cells, first with a competition binding assay then with a chemotaxis assay along a C-X-C motif chemokine ligand 12 (CXCL12) concentration gradient. At 20 μM, the bis(cyclam)-capped cholesterol lipid was as effective as the commercial drug AMD3100 for preventing the migration of CLL cells, despite a lower affinity for CXCR4 than AMD3100.
Sigma-Aldrich
Cholesteryl chloroformate, 95%
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)