MilliporeSigma
  • Scale-invariant patterning by size-dependent inhibition of Nodal signalling.
Scale-invariant patterning by size-dependent inhibition of Nodal signalling.
Nature cell biology (2018-08-01)
María Almuedo-Castillo, Alexander Bläßle, David Mörsdorf, Luciano Marcon, Gary H Soh, Katherine W Rogers, Alexander F Schier, Patrick Müller
ABSTRACT
Individuals can vary substantially in size, but the proportions of their body plans are often maintained. We generated smaller zebrafish by removing 30% of their cells at the blastula stages and found that these embryos developed into normally patterned individuals. Strikingly, the proportions of all germ layers adjusted to the new embryo size within 2 hours after cell removal. As Nodal-Lefty signalling controls germ-layer patterning, we performed a computational screen for scale-invariant models of this activator-inhibitor system. This analysis predicted that the concentration of the highly diffusive inhibitor Lefty increases in smaller embryos, leading to a decreased Nodal activity range and contracted germ-layer dimensions. In vivo studies confirmed that Lefty concentration increased in smaller embryos, and embryos with reduced Lefty levels or with diffusion-hindered Lefty failed to scale their tissue proportions. These results reveal that size-dependent inhibition of Nodal signalling allows scale-invariant patterning.
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep