MilliporeSigma
  • Metal-organic framework-derived mesoporous octahedral copper oxide/titania composites for high-performance lithium-ion batteries.
Metal-organic framework-derived mesoporous octahedral copper oxide/titania composites for high-performance lithium-ion batteries.
Journal of colloid and interface science (2018-06-17)
Dan Ping Wang, Maosen Fu, Yuan Ha, Hao Wang, Renbing Wu
ABSTRACT
A mesoporous octahedral copper oxide@titania (CuO/TiO2) composites with core-shelled structure have been successfully fabricated via a facile and cost-effective approach, which involves two main steps: the creation of homogeneous TiO2 shell onto the octahedral Cu-based metal-organic frameworks (MOFs) template and thermal decomposition of the template at controlled temperature in the air. The design of combining CuO with the TiO2 layer within a porous octahedra structure is beneficial to integrate the advantages of different components and address the severe volume change associated with pulverization issue that exists in most metal oxides-based electrodes. When assembled as an anode material for lithium-ion batteries, the as-fabricated mesoporous CuO/TiO2 octahedra can achieve outstanding electrochemical performance in terms of a high reversible capacity (692 mAh g-1 at 100 mA g-1 for over 200 cycles) and exceptional rate capability (441 and 387 mA h g-1 at 1600 and 3200 mA g-1, respectively).