MilliporeSigma
  • Functional and structural biomarkers to monitor heavy metal pollution of one of the most contaminated freshwater sites in Southern Europe.
Functional and structural biomarkers to monitor heavy metal pollution of one of the most contaminated freshwater sites in Southern Europe.
Ecotoxicology and environmental safety (2018-08-12)
Viviana Maresca, Lina Fusaro, Sergio Sorbo, Antonietta Siciliano, Stefano Loppi, Luca Paoli, Fabrizio Monaci, Elham Asadi Karam, Marina Piscopo, Marco Guida, Emilia Galdiero, Marilena Insolvibile, Adriana Basile
ABSTRACT
This study evaluated the biological effects of highly polluted freshwater environment (Regi Lagni channels, S Italy) on the aquatic moss Leptodictyum riparium, exposed in bags at three sites representative of different environmental conditions and characterized by different heavy metal burdens. Bioaccumulation, ultrastructural alterations, Reactive Oxygen Species (ROS) production, antioxidant enzymes activity and DNA damage were assessed. To better evaluate the biological response of the moss species to heavy metals, the same biological parameters were assessed also in L. riparium samples cultured in vitro using metal mixtures at the same concentrations as measured at the 3 field exposure sites. Heavy metals were accumulated into the moss tissues causing severe ultra-structural damages at higher concentration case studies, and the ROS production as well as the activity of the enzyme followed a concentration-dependent increase. However, the DNA damage trend suggested a threshold effect that changed between field and in vitro experiment. The enrichment factor suggests that the concentration in the most polluted site is close to the upper limit of L. riparium to accumulate metals. Overall, combining measures of the morpho-functional traits at different level contribute to improving the knowledge about the tolerance of L. riparium to heavy metal stress, suggesting that this moss could be suitable for biomonitoring activity in field conditions.
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests