MilliporeSigma
  • Isolation and differential transcriptome of vascular smooth muscle cells and mid-capillary pericytes from the rat brain.
Isolation and differential transcriptome of vascular smooth muscle cells and mid-capillary pericytes from the rat brain.
Scientific reports (2018-08-18)
Stéphanie Chasseigneaux, Yasmine Moraca, Véronique Cochois-Guégan, Anne-Cécile Boulay, Alice Gilbert, Stéphane Le Crom, Corinne Blugeon, Cyril Firmo, Salvatore Cisternino, Jean-Louis Laplanche, Emmanuel Curis, Xavier Declèves, Bruno Saubaméa
ABSTRACT
Brain mural cells form a heterogeneous family which significantly contributes to the maintenance of the blood-brain barrier and regulation of the cerebral blood flow. Current procedures to isolate them cannot specifically separate their distinct subtypes, in particular vascular smooth muscle cells (VSMCs) and mid-capillary pericytes (mcPCs), which differ among others by their expression of smooth muscle actin (SMA). We herein describe an innovative method allowing SMA+ VSMCs and SMA- mcPCs to be freshly isolated from the rat cerebral cortex. Using differential RNA-Seq analysis, we then reveal the specific gene expression profile of each subtype. Our results refine the current description of the role of VSMCs in parenchymal cortical arterioles at the molecular level and provide a unique platform to identify the molecular mechanisms underlying the specific functions of mcPCs in the brain vasculature.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%