MilliporeSigma
Discovery of a PCAF Bromodomain Chemical Probe.
Angewandte Chemie (International ed. in English) (2016-12-15)
Moses Moustakim, Peter G K Clark, Laura Trulli, Angel L Fuentes de Arriba, Matthias T Ehebauer, Apirat Chaikuad, Emma J Murphy, Jacqui Mendez-Johnson, Danette Daniels, Chun-Feng D Hou, Yu-Hui Lin, John R Walker, Raymond Hui, Hongbing Yang, Lucy Dorrell, Catherine M Rogers, Octovia P Monteiro, Oleg Fedorov, Kilian V M Huber, Stefan Knapp, Jag Heer, Darren J Dixon, Paul E Brennan
ABSTRACT
The p300/CBP-associated factor (PCAF) and related GCN5 bromodomain-containing lysine acetyl transferases are members of subfamily I of the bromodomain phylogenetic tree. Iterative cycles of rational inhibitor design and biophysical characterization led to the discovery of the triazolopthalazine-based L-45 (dubbed L-Moses) as the first potent, selective, and cell-active PCAF bromodomain (Brd) inhibitor. Synthesis from readily available (1R,2S)-(-)-norephedrine furnished L-45 in enantiopure form. L-45 was shown to disrupt PCAF-Brd histone H3.3 interaction in cells using a nanoBRET assay, and a co-crystal structure of L-45 with the homologous Brd PfGCN5 from Plasmodium falciparum rationalizes the high selectivity for PCAF and GCN5 bromodomains. Compound L-45 shows no observable cytotoxicity in peripheral blood mononuclear cells (PBMC), good cell-permeability, and metabolic stability in human and mouse liver microsomes, supporting its potential for in vivo use.