MilliporeSigma
  • Magnetic-assisted biotinylated single-chain variable fragment antibody-based immunoassay for amantadine detection in chicken.
Magnetic-assisted biotinylated single-chain variable fragment antibody-based immunoassay for amantadine detection in chicken.
Analytical and bioanalytical chemistry (2018-07-15)
Sanlei Xie, Kai Wen, Jie Xie, Yongjun Zheng, Tao Peng, Jianyi Wang, Kai Yao, Shuangyang Ding, Haiyang Jiang
ABSTRACT
A sensitive competitive immunoassay with simple operation was developed for the detection of the anti-virus drug amantadine (AMD). The single-chain variable fragment (scFv) antibody against AMD was site-specific biotinylated and overexpressed as a secreted body in Escherichia coli AVB101. Horseradish peroxidase-labeled streptavidin-biotinylated scFv antibody (HRP-SA-BIO-scFv) could specifically bind to AMD-functionalized magnetic beads (MBs) and then the immune complexes were separated from the matrix solution by magnet. The concentration of the AMD could be known by the measurement of the signal produced by the horseradish peroxidase. The newly established assay provides a significant improvement in comparison to the conventional ELISA without SA-BIO signal amplification and MBs separation. The limit of detection and assay time was 0.64 vs. 8.4 ng/mL and 50 vs. 150 min, respectively. The recoveries ranged from 77.8 to 112% with the coefficient of variation less than 13%. The immunoassay exhibited an obvious cross-reactivity to rimantadine (84%), 1-(1-adamantyl)ethylamine (72%), and somantadine (63%). These results demonstrated that the developed immunoassay provided a sensitive, rapid, and accurate approach for the detection of AMD in chicken by employing MBs as solid phase and SA-BIO as signal amplification. When applied in natural chicken samples, the newly established method provided results consistent with those from UPLC-MS/MS, suggesting that the proposed method could be used for rapid screening of the target of interest; the new immunoassay could also be extended to other small molecular contaminants and thus represents a universal strategy for food safety analysis. Graphical abstract ᅟ.
Sigma-Aldrich
2-Adamantanol, 97%
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)