MilliporeSigma
  • TORC-SIK cascade regulates CREB activity through the basic leucine zipper domain.
TORC-SIK cascade regulates CREB activity through the basic leucine zipper domain.
The FEBS journal (2007-06-15)
Hiroshi Takemori, Junko Kajimura, Mitsuhiro Okamoto
ABSTRACT
The transcription factor cAMP response element-binding protein (CREB) plays important roles in gene expression induced by cAMP signaling and is believed to be activated when its Ser133 is phosphorylated. However, the discovery of Ser133-independent activation by the activation of transducer of regulated CREB activity coactivators (TORC) and repression by salt inducible kinase cascades suggests that Ser133-independent regulation of CREB is also important. The activation and repression are mediated by the basic leucine zipper domain of CREB. In this review, we focus on the basic leucine zipper domain in the regulation of transcriptional activity of CREB and describe the functions of TORC and salt inducible kinase.
Sigma-Aldrich
SIK (1-303), active, His tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution