MilliporeSigma
NAK is an IkappaB kinase-activating kinase.
Nature (2000-04-28)
Y Tojima, A Fujimoto, M Delhase, Y Chen, S Hatakeyama, K Nakayama, Y Kaneko, Y Nimura, N Motoyama, K Ikeda, M Karin, M Nakanishi
ABSTRACT
Phosphorylation of IkappaB by the IkappaB kinase (IKK) complex is a critical step leading to IkappaB degradation and activation of transcription factor NF-kappaB. The IKK complex contains two catalytic subunits, IKKalpha and IKKbeta, the latter being indispensable for NF-kappaB activation by pro-inflammatory cytokines. Although IKK is activated by phosphorylation of the IKKbeta activation loop, the physiological IKK kinases that mediate responses to extracellular stimuli remain obscure. Here we describe an IKK-related kinase, named NAK (NF-kappaB-activating kinase), that can activate IKK through direct phosphorylation. NAK induces IkappaB degradation and NF-kappaB activity through IKKbeta. Endogenous NAK is activated by phorbol ester tumour promoters and growth factors, whereas catalytically inactive NAK specifically inhibits activation of NF-kappaB by protein kinase C-epsilon (PKCepsilon). Thus, NAK is an IKK kinase that may mediate IKK and NF-kappaB activation in response to growth factors that stimulate PKCepsilon activity.
Sigma-Aldrich
TBK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution