MilliporeSigma
  • Cascade reaction of isatins with heterocyclic ketene aminals: synthesis of imidazopyrroloquinoline derivatives.
Cascade reaction of isatins with heterocyclic ketene aminals: synthesis of imidazopyrroloquinoline derivatives.
Organic letters (2011-08-19)
Fuchao Yu, Shengjiao Yan, Ling Hu, Yongchao Wang, Jun Lin
ABSTRACT
A concise and efficient route for the synthesis of highly substituted imidazopyrroloquinoline derivatives by simply refluxing a reaction mixture of different types of isatins and heterocyclic ketene aminals (HKAs) by acetic acid was developed. This method is suitable for combinatorial and parallel syntheses in drug discovery; consequently, a library of highly substituted imidazopyrroloquinoline derivatives was rapidly constructed using the present protocol.
Sigma-Aldrich
Isatin, 97%