MilliporeSigma
  • Ankyrin repeat and zinc-finger domain-containing 1 mutations are associated with infantile-onset inflammatory bowel disease.
Ankyrin repeat and zinc-finger domain-containing 1 mutations are associated with infantile-onset inflammatory bowel disease.
The Journal of biological chemistry (2017-03-18)
Désirée Y van Haaften-Visser, Magdalena Harakalova, Enric Mocholi, Joris M van Montfrans, Abdul Elkadri, Ester Rieter, Karoline Fiedler, Peter M van Hasselt, Emily M M Triffaux, Mieke M van Haelst, Isaac J Nijman, Wigard P Kloosterman, Edward E S Nieuwenhuis, Aleixo M Muise, Edwin Cuppen, Roderick H J Houwen, Paul J Coffer
ABSTRACT
Infantile-onset inflammatory bowel disease (IO IBD) is an invalidating illness with an onset before 2 years of age and has a complex pathophysiology in which genetic factors are important. Homozygosity mapping and whole-exome sequencing in an IO IBD patient and subsequent sequencing of the candidate gene in 12 additional IO IBD patients revealed two patients with two mutated ankyrin repeat and zinc-finger domain-containing 1 (ANKZF1) alleles (homozygous ANKZF1 R585Q mutation and compound heterozygous ANKZF1 E152K and V32_Q87del mutations, respectively) and two patients with one mutated ANKZF1 allele. Although the function of ANKZF1 in mammals had not been previously evaluated, we show that ANKZF1 has an indispensable role in the mitochondrial response to cellular stress. ANKZF1 is located diffusely in the cytoplasm and translocates to the mitochondria upon cellular stress. ANKZF1 depletion reduces mitochondrial integrity and mitochondrial respiration under conditions of cellular stress. The ANKZF1 mutations identified in IO IBD patients with two mutated ANKZF1 alleles result in dysfunctional ANKZF1, as shown by an increased level of apoptosis in patients' lymphocytes, a decrease in mitochondrial respiration in patient fibroblasts with a homozygous ANKZF1 R585Q mutation, and an inability of ANKZF1 R585Q and E152K to rescue the phenotype of yeast deficient in Vms1, the yeast homologue of ANKZF1. These data indicate that loss-of-function mutations in ANKZF1 result in deregulation of mitochondrial integrity, and this may play a pathogenic role in the development of IO IBD.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ANKZF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit