MilliporeSigma
  • Synthesis of P2C2O2 and P2CO via NHC-mediated coupling of the phosphaethynolate anion.
Synthesis of P2C2O2 and P2CO via NHC-mediated coupling of the phosphaethynolate anion.
Chemical communications (Cambridge, England) (2017-11-03)
Robert J Gilliard, Riccardo Suter, Erik Schrader, Zoltán Benkő, Arnold L Rheingold, Hansjörg Grützmacher, John D Protasiewicz
ABSTRACT
The reaction of the chloroimidazolium chloride salt, [NHC-Cl][Cl], NHC = C{N(2,6-iPr2C6H3)CH}2 (1) with two equivalents of sodium phosphaethynolate, Na[OCP]·(dioxane)2.5, results in the formation of NHC-{cyclo-(CO)-P2-C(O)} (2) and NHC-P2-C(O)-NHC (3). Notably, in the presence of free NHC ligand, compound 2 converts to compound 3via extrusion of CO at elevated temperatures. The nature of the bonding in these complexes was probed computationally and spectroscopically.
Sigma-Aldrich
2-Chloro-1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, ≥95%
Pricing and availability is not currently available.