MilliporeSigma
Proteasome inhibition by peptide-semicarbazones.
Bioorganic & medicinal chemistry (2008-03-04)
Johann Leban, Marcus Blisse, Babett Krauss, Sandra Rath, Roland Baumgartner, Markus H J Seifert
ABSTRACT
Peptide-semicarbazones derived from Z-Trp-Trp-Phe-aldehyde inhibit the chymotryptic activity of the human proteasome at nanomolar concentrations, but are less active in a NFkappaB reporter gene assay. Cyclic semicarbazones, in contrast, combine a strong inhibitory effect on the enzyme with an inhibition of NFkappaB signaling in the nanomolar range. In addition, a practical synthesis for scale-up of such compounds was developed.
Sigma-Aldrich
1-Aminohydantoin hydrochloride, 98%