MilliporeSigma
CRISPR-Cas9 genome editing induces a p53-mediated DNA damage response.
Nature medicine (2018-06-13)
Emma Haapaniemi, Sandeep Botla, Jenna Persson, Bernhard Schmierer, Jussi Taipale
ABSTRACT
Here, we report that genome editing by CRISPR-Cas9 induces a p53-mediated DNA damage response and cell cycle arrest in immortalized human retinal pigment epithelial cells, leading to a selection against cells with a functional p53 pathway. Inhibition of p53 prevents the damage response and increases the rate of homologous recombination from a donor template. These results suggest that p53 inhibition may improve the efficiency of genome editing of untransformed cells and that p53 function should be monitored when developing cell-based therapies utilizing CRISPR-Cas9.
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)