MilliporeSigma
  • MRI Is a DNA Damage Response Adaptor during Classical Non-homologous End Joining.
MRI Is a DNA Damage Response Adaptor during Classical Non-homologous End Joining.
Molecular cell (2018-07-19)
Putzer J Hung, Britney Johnson, Bo-Ruei Chen, Andrea K Byrum, Andrea L Bredemeyer, William T Yewdell, Tanya E Johnson, Brian J Lee, Shruthi Deivasigamani, Issa Hindi, Parmeshwar Amatya, Michael L Gross, Tanya T Paull, David J Pisapia, Jayanta Chaudhuri, John J H Petrini, Nima Mosammaparast, Gaya K Amarasinghe, Shan Zha, Jessica K Tyler, Barry P Sleckman
ABSTRACT
The modulator of retrovirus infection (MRI or CYREN) is a 30-kDa protein with a conserved N-terminal Ku-binding motif (KBM) and a C-terminal XLF-like motif (XLM). We show that MRI is intrinsically disordered and interacts with many DNA damage response (DDR) proteins, including the kinases ataxia telangiectasia mutated (ATM) and DNA-PKcs and the classical non-homologous end joining (cNHEJ) factors Ku70, Ku80, XRCC4, XLF, PAXX, and XRCC4. MRI forms large multimeric complexes that depend on its N and C termini and localizes to DNA double-strand breaks (DSBs), where it promotes the retention of DDR factors. Mice deficient in MRI and XLF exhibit embryonic lethality at a stage similar to those deficient in the core cNHEJ factors XRCC4 or DNA ligase IV. Moreover, MRI is required for cNHEJ-mediated DSB repair in XLF-deficient lymphocytes. We propose that MRI is an adaptor that, through multivalent interactions, increases the avidity of DDR factors to DSB-associated chromatin to promote cNHEJ.
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
Sigma-Aldrich
Anti-ATRIP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Imatinib
Millipore
EZview Red Anti-HA Affinity Gel
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution