MilliporeSigma
  • Inhibitory effect of curcuminoid pretreatments on endogenous dentin proteases.
Inhibitory effect of curcuminoid pretreatments on endogenous dentin proteases.
Dental materials journal (2018-03-02)
Roda Seseogullari-Dirihan, Meltem Tekbas Atay, David Henry Pashley, Arzu Tezvergil-Mutluay
ABSTRACT
The aim of this study was to evaluate the effect of curcuminoids on the dentin endogenous protease activity. Demineralized dentin were pretreated with 50 or 100 µM of three different curcuminoids for 60 s and incubated up to 3 months. Untreated beams served as controls. Dry dentin mass was measured after incubation. Aliquots were analyzed for the quantity of ICTP and CTX releases for MMP and cathepsin-K mediated degradation, respectively. The effect of curcuminoids on matrix-bound MMP and soluble rhMMP-9 were measured using an activity assay. Data were subjected to repeated-measures-ANOVA (α=0.05). Gelatinolytic activity was analyzed using zymography. ICTP and CTX release and dry mass loss of curcuminoid-treated groups were significantly lower than the control. Inhibition of rhMMP-9 varied from 29-49% among curcumonoid-treated groups, whereas no inhibition was observed at untreated control (p>0.05). Results were confirmed by zymography. The study showed that the pretreatment of dentin matrices by curcuminoids decreases endogenous protease activity-mediated degradation in dentin.