MilliporeSigma
  • Basal level of autophagy and MAP1LC3B-II as potential biomarkers for DHA-induced cytotoxicity in colorectal cancer cells.
Basal level of autophagy and MAP1LC3B-II as potential biomarkers for DHA-induced cytotoxicity in colorectal cancer cells.
The FEBS journal (2018-05-04)
Helle Samdal, Malin A Sandmoe, Lene C Olsen, Elaf A H Jarallah, Therese S Høiem, Svanhild A Schønberg, Caroline H H Pettersen
ABSTRACT
The omega-3 fatty acid docosahexaenoic acid (DHA) is known as an anticancer agent. Colorectal cancer (CRC) cells exhibit different sensitivity toward DHA, but the mechanisms involved are still unclear. Gene expression profiling of 10 CRC cell lines demonstrated a correlation between the level of DHA sensitivity and different biological stress responses, such as endoplasmic reticulum (ER) stress, oxidative stress, and autophagy. The basal level of autophagy and MAP1LC3B-II protein correlated with DHA sensitivity in the cell lines studied. DHA induced oxidative stress, ER stress, and autophagy in DHA-sensitive DLD-1 cells, while the less sensitive LS411N cells were affected to a much lesser extent. Co-treatment with DHA and the autophagy inducer rapamycin reduced DHA sensitivity in DLD-1 and HCT-8 cells, while co-treatment with DHA and the autophagy inhibitors chloroquine and 3-methyladenine increased the DHA sensitivity in LS411N and LS513 cells. Differentially expressed genes correlating with DHA sensitivity and the level of autophagy demonstrated an overlap in biological pathways involved. Results indicate the basal level of autophagy and MAP1LC3B-II protein as potential biomarkers for DHA sensitivity in CRC cells. Protocol and data for gene expression experiments have been submitted to ArrayExpress with accession number E-MTAB-5750.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53