MilliporeSigma
  • Matrine suppresses KRAS-driven pancreatic cancer growth by inhibiting autophagy-mediated energy metabolism.
Matrine suppresses KRAS-driven pancreatic cancer growth by inhibiting autophagy-mediated energy metabolism.
Molecular oncology (2018-05-24)
Young-Ra Cho, Ji Hyeon Lee, Ji Hye Kim, So-Yeon Lee, Suna Yoo, Min-Kyo Jung, Su Jung Kim, Hyun Ju Yoo, Chang-Gi Pack, Jin Kyung Rho, Jaekyoung Son
ABSTRACT
Matrine is a natural compound extracted from the herb Sophora flavescens Ait which is widely used in traditional Chinese medicine for treating various diseases. Recently, matrine was reported to have antitumor effects against a variety of cancers without any obvious side effects; however, the molecular mechanisms of its antiproliferative effects on cancer are unclear. Here, we report that matrine inhibits autophagy-mediated energy metabolism, which is necessary for pancreatic cancer growth. We found that matrine significantly reduces pancreatic cancer growth in vitro and in vivo by insufficiently maintaining mitochondrial metabolic function and energy level. We also found that either pyruvate or α-ketoglutarate supplementation markedly rescues pancreatic cancer cell growth following matrine treatment. Inhibition of mitochondrial energy production results from matrine-mediated autophagy inhibition by impairing the function of lysosomal protease. Matrine-mediated autophagy inhibition requires stat3 downregulation. Furthermore, we found that the antitumor effect of matrine on pancreatic cancer growth depends on the mutation of the KRAS oncogene. Together, our data suggest that matrine can suppress the growth of KRAS-mutant pancreatic cancer by inhibiting autophagy-mediated energy metabolism.
Sigma-Aldrich
Methyl pyruvate, 90%, technical grade