MilliporeSigma
  • A highly versatile catalyst system for the cross-coupling of aryl chlorides and amines.
A highly versatile catalyst system for the cross-coupling of aryl chlorides and amines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-12-22)
Rylan J Lundgren, Antonia Sappong-Kumankumah, Mark Stradiotto
ABSTRACT
The syntheses of 2-(di-tert-butylphosphino)-N,N-dimethylaniline (L1, 71%) and 2-(di-1-adamantylphosphino)-N,N-dimethylaniline (L2, 74 %), and their application in Buchwald-Hartwig amination, are reported. In combination with [Pd(allyl)Cl](2) or [Pd(cinnamyl)Cl](2), these structurally simple and air-stable P,N ligands enable the cross-coupling of aryl and heteroaryl chlorides, including those bearing as substituents enolizable ketones, ethers, esters, carboxylic acids, phenols, alcohols, olefins, amides, and halogens, to a diverse range of amine and related substrates that includes primary alkyl- and arylamines, cyclic and acyclic secondary amines, N-H imines, hydrazones, lithium amide, and ammonia. In many cases, the reactions can be performed at low catalyst loadings (0.5-0.02 mol % Pd) with excellent functional group tolerance and chemoselectivity. Examples of cross-coupling reactions involving 1,4-bromochlorobenzene and iodobenzene are also reported. Under similar conditions, inferior catalytic performance was achieved when using Pd(OAc)(2), PdCl(2), [PdCl(2)(cod)] (cod = 1,5-cyclooctadiene), [PdCl(2)(MeCN)(2)], or [Pd(2)(dba)(3)] (dba = dibenzylideneacetone) in combination with L1 or L2, or by use of [Pd(allyl)Cl](2) or [Pd(cinnamyl)Cl](2) with variants of L1 and L2 bearing less basic or less sterically demanding substituents on phosphorus or lacking an ortho-dimethylamino fragment. Given current limitations associated with established ligand classes with regard to maintaining high activity across the diverse possible range of C-N coupling applications, L1 and L2 represent unusually versatile ligand systems for the cross-coupling of aryl chlorides and amines.
Sigma-Aldrich
Lithium amide, powder, 95%