MilliporeSigma
  • Pontamine sky blue: a counterstain for background autofluorescence in fluorescence and immunofluorescence histochemistry.
Pontamine sky blue: a counterstain for background autofluorescence in fluorescence and immunofluorescence histochemistry.
Histochemistry (1985-01-01)
T Cowen, A J Haven, G Burnstock
ABSTRACT
The stain pontamine sky blue (PSB) has been shown to reduce background autofluorescence in catecholamine fluorescence and immunofluorescence histochemical preparations. Using PSB as a counterstain on whole-mount stretch preparations of human mesenteric blood vessels, a medium dense noradrenergic nerve plexus is clearly revealed, which previously had been only partially visible because of background autofluorescence. Image analysis of nerve densities in whole-mount stretch preparations of guinea-pig arteries containing noradrenergic, substance P-, and vasoactive intestinal polypeptide (VIP)-positive nerve plexuses shows that PSB staining does not alter the specific neuronal fluorescence and that it improves image definition.
Sigma-Aldrich
Chicago Sky Blue 6B, powder