MilliporeSigma
  • Dual Requirement of CHD8 for Chromatin Landscape Establishment and Histone Methyltransferase Recruitment to Promote CNS Myelination and Repair.
Dual Requirement of CHD8 for Chromatin Landscape Establishment and Histone Methyltransferase Recruitment to Promote CNS Myelination and Repair.
Developmental cell (2018-06-20)
Chuntao Zhao, Chen Dong, Magali Frah, Yaqi Deng, Corentine Marie, Feng Zhang, Lingli Xu, Zhixing Ma, Xinran Dong, Yifeng Lin, Scott Koenig, Brahim Nait-Oumesmar, Donna M Martin, Laiman N Wu, Mei Xin, Wenhao Zhou, Carlos Parras, Q Richard Lu
ABSTRACT
Disruptive mutations in chromatin remodeler CHD8 cause autism spectrum disorders, exhibiting widespread white matter abnormalities; however, the underlying mechanisms remain elusive. We show that cell-type specific Chd8 deletion in oligodendrocyte progenitors, but not in neurons, results in myelination defects, revealing a cell-intrinsic dependence on CHD8 for oligodendrocyte lineage development, myelination and post-injury remyelination. CHD8 activates expression of BRG1-associated SWI/SNF complexes that in turn activate CHD7, thus initiating a successive chromatin remodeling cascade that orchestrates oligodendrocyte lineage progression. Genomic occupancy analyses reveal that CHD8 establishes an accessible chromatin landscape, and recruits MLL/KMT2 histone methyltransferase complexes distinctively around proximal promoters to promote oligodendrocyte differentiation. Inhibition of histone demethylase activity partially rescues myelination defects of CHD8-deficient mutants. Our data indicate that CHD8 exhibits a dual function through inducing a cascade of chromatin reprogramming and recruiting H3K4 histone methyltransferases to establish oligodendrocyte identity, suggesting potential strategies of therapeutic intervention for CHD8-associated white matter defects.
Sigma-Aldrich
Anti-NOGO A Antibody, Chemicon®, from rabbit
Sigma-Aldrich
MISSION® esiRNA, targeting human CHD8
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-O4 Antibody, clone 81, clone 81 (mAB O4), Chemicon®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human WDR5
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep