MilliporeSigma
  • Metronomic capecitabine vs. best supportive care in Child-Pugh B hepatocellular carcinoma: a proof of concept.
Metronomic capecitabine vs. best supportive care in Child-Pugh B hepatocellular carcinoma: a proof of concept.
Scientific reports (2018-07-04)
Stefania De Lorenzo, Francesco Tovoli, Maria Aurelia Barbera, Francesca Garuti, Andrea Palloni, Giorgio Frega, Ingrid Garajovà, Alessandro Rizzo, Franco Trevisani, Giovanni Brandi
ABSTRACT
There is a relative lack of evidence about systemic treatments in patients with hepatocellular carcinoma (HCC) and moderate liver dysfunction (Child-Pugh B). In this multicenter study we retrospectively analyzed data from Child-Pugh B-HCC patients naïve to systemic therapies, treated with MC or best supportive care (BSC). To reduce the risk of selection bias, an inverse probability of treatment weighting approach was adopted. Propensity score was generated including: extrahepatic spread; macrovascular invasion; performance status, alphafetoprotein > 400 ng/ml, Child- Pugh score [B7 vs. B8-9]. We identified 35 MC-treated patients and 70 controls. Median overall survival was 7.5 [95% CI: 3.733-11.267]in MC-patients and 5.1 months [95% CI: 4.098-6.102] in the BSC group (p = 0.013). In patients treated with MC, median progression-free survival was 4.5 months (95% CI: 2.5-6.5). The univariate unweighted Cox regression showed a 42% reduction in death risk for patients on MC (95%CI: 0.370-0.906; p = 0.017). After weighting for potential confounders, death risk remained essentially unaltered. In the MC group, 12 patients (34.3%) experienced at least one adverse event, the most common of which were: fatigue (17.1%), hand-foot syndrome (8.5%), thrombocytopenia (8.5%), and neutropenia (5.7%). MC seems a safe option for Child-Pugh B-HCC patients. Its potential antitumour activity warrants prospective evaluations.
Sigma-Aldrich
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)