MilliporeSigma
  • Comparative Bio-Efficacy and Synergism of New Generation Polyfluorobenzyl and Conventional Pyrethroids Against Culex quinquefasciatus (Diptera: Culicidae).
Comparative Bio-Efficacy and Synergism of New Generation Polyfluorobenzyl and Conventional Pyrethroids Against Culex quinquefasciatus (Diptera: Culicidae).
Journal of economic entomology (2018-04-17)
Manas Sarkar, Ambadas Akulwad, Rajendra Kshirsagar, Siva Muthukrishnan
ABSTRACT
Intensive exposure to insecticides has resulted in the evolution of insecticide resistance in the mosquitoes. We tested the bio-efficacy of two Culex quinquefasciatus Say (Diptera: Culicidae) laboratory strains differentially bio-responsive to pyrethroids to understand the comparative efficacy of different polyfluorobenzyle and conventional pyrethroid molecules and the role of piperonyl butoxide (PBO) in synergizing these molecules in increased tolerance of mosquitoes to these molecules. We have taken deltamethrin (α-cyano pyrethroid with phenoxybenzyl moiety); permethrin (phenoxybenzyl pyrethroid without an α-cyano group); transfluthrin, dimefluthrin, metofluthrin, and meperfluthrin (polyfluorinated benzyl compounds); and prallethrin (modified cyclopentadienone compound) for this study. We found higher bio-efficacy in dimefluthrin, metofluthrin, and meperfluthrin compared with transfluthrin against tested mosquito strains. We found that transfluthrin exhibited synergism with PBO, which supports the hypothesis that P450 enzymes could play a role in the detoxification process of transfluthrin, which was earlier not believed. However, other polyfluorobenzyl pyrethroids with a 4-(methoxymethyl) phenyl capping in the tetrafluorobenzyl ring (dimefluthrin, metofluthrin, and meperfluthrin) exhibit greater synergism with PBO compared with transfluthrin. Further study is required to understand the mechanism for higher synergistic ratios in polyfluorobenzyl pyrethroids with 4-(methoxymethyl) phenyl moiety and ascertain the possible involvement of novel mechanisms that may involve in developing resistance. This is the first report of comparative bio-efficacy of multiple polyfluorobenzyl pyrethroids and PBO synergism against mosquitoes.
Product Description
Supelco
Transfluthrin, PESTANAL®, analytical standard
Supelco
Prallethrin, PESTANAL®, analytical standard