MilliporeSigma
Magnetic field-directed hybrid anisotropic nanocomposites.
Nanotechnology (2018-06-05)
Maogang Gong, Jingming Zhang, Shenqiang Ren
ABSTRACT
A facile bottom-up approach is developed to grow magnetic metallic Cu/FeCo (core/shell) nanowires, where their distribution and orientation can be controlled by magnetic field. The nanocomposites consisting of a ferroelectric polymer matrix and magnetic nanowire arrays exhibit the orientation-controlled anisotropy and interfacial magnetoelectric coupling effect.
Sigma-Aldrich
Nickel(II) acetylacetonate, 95%
Sigma-Aldrich
DCB, ≥98% (HPLC), solid