MilliporeSigma
  • Implementation of a generic liquid chromatographic method development workflow: Application to the analysis of phytocannabinoids and Cannabis sativa extracts.
Implementation of a generic liquid chromatographic method development workflow: Application to the analysis of phytocannabinoids and Cannabis sativa extracts.
Journal of pharmaceutical and biomedical analysis (2018-04-09)
Szabolcs Fekete, Vida Sadat-Noorbakhsh, Cedric Schelling, Imre Molnár, Davy Guillarme, Serge Rudaz, Jean-Luc Veuthey
ABSTRACT
A generic liquid chromatographic method development workflow was developed and successfully applied to the analysis of phytocannabinoids and Cannabis sativa extracts. Our method development procedure consists in four steps: At the end, this method development workflow was performed in less than 4 days and minimizes the costs of the method development in liquid chromatography.