MilliporeSigma
  • Repurposing itraconazole to the benefit of skin cancer treatment: A combined azole-DDAB nanoencapsulation strategy.
Repurposing itraconazole to the benefit of skin cancer treatment: A combined azole-DDAB nanoencapsulation strategy.
Colloids and surfaces. B, Biointerfaces (2018-04-24)
C Carbone, C Martins-Gomes, V Pepe, A M Silva, T Musumeci, G Puglisi, P M Furneri, E B Souto
ABSTRACT
In this work, we aimed at developing an improved topical SLN formulation combining itraconazole delivery with a coating layer of didodecyldimethylammonium bromide, thus repurposing the drug effectiveness by synergistic skin anticancer effectiveness. In order to obtain a stable SLN formulation with small homogeneously dispersed particles, a deep formulative study was developed screening three different solid lipids (Suppocire NB, Cetyl Palmitate and Dynasan 114) for the SLN preparation by the phase inversion temperature. A bluishcolored shade formulation, with homogeneous small particles size (<50 nm) was obtained only using Suppocire NB. The cytotoxicity of all SLN was tested after 24 h exposure against three adherent skin cell lines (A431, HaCaT and SK-MEL-5). Results demonstrate that both unloaded and drugloaded SLN did not significantly affect the cell viability of the non-tumoral HaCaT cell line, thus confirming the safe potential topical application of these formulations. A dose-dependent decrease in cell viability was observed for the tumoral cell lines, A431 and SK-MEL-5, with a significant reduction of the A431 cancer cell line viability. The drug molecule addition to the uncoated nanoparticles was able to increase of almost 20% the reduction of the viability of the cancer cells treated. Ours results demonstrate the potentiality of repurposing itraconazole activity by using the combined nanoencapsulation strategy with the positively charged coating layer on SLN, which can be further investigated as a promising stable and safe approach to significantly reduce the viability of skin cancer cells.
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%