MilliporeSigma
  • Delivery of oat-derived phytoceramides into the stratum corneum of the skin using nanocarriers: Formulation, characterization and in vitro and ex-vivo penetration studies.
Delivery of oat-derived phytoceramides into the stratum corneum of the skin using nanocarriers: Formulation, characterization and in vitro and ex-vivo penetration studies.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2018-03-05)
Efrem N Tessema, Tsige Gebre-Mariam, Getahun Paulos, Johannes Wohlrab, Reinhard H H Neubert
ABSTRACT
Deficiency or altered composition of stratum corneum (SC) lipids such as ceramides (CERs), causing skin barrier dysfunction and skin dryness, have been associated with skin diseases such as atopic dermatitis and psoriasis, and ageing. Replenishing the depleted native CERs with exogenous CERs has also been shown to have beneficial effects in restoring the skin barrier. Phyto-derived CERs such as oat CERs were shown to be potential for skin barrier reinforcement. To effect this, however, the oat CERs should overcome the SC barrier and delivered deep into the lipid matrix using the various novel formulations. In an attempt to demonstrate the potential use of oat CERs, lecithin-based microemulsions (MEs) and starch-based nanoparticles (NPs) were formulated and characterized. Besides, ME gel and NP gel were also prepared using Carbopol®980 as a gelling agent. The in vitro release and penetration (using artificial four-layer membrane system) and ex vivo permeation (using excised human skin) of oat CERs from the various formulations were investigated. The results revealed ME enhanced the in vitro release and penetration oat CERs compared to the other formulations. On the other hand, the NPs retarded the release of oat CERs and small quantities of oat CERs incorporated into NP gel penetrated into the deeper layers of the multilayer membranes. The penetration-enhancing effect of ME was also observed in the ex vivo permeation studies where significant quantities of oat CERs were found in the acceptor compartment. Compared to the ME, the ME gel exhibited reduced depth and extent of oat CERs permeation. As compared to NP gel, ME gel enhanced the degree of permeation of oat CERs into the deeper layer of the skin. Generally the gel formulations were effective in concentrating oat CERs in the SC where they are needed to be.
Sigma-Aldrich
1,2-Pentanediol, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Pricing and availability is not currently available.