MilliporeSigma
  • Effect of Presence and Concentration of Plasticizers, Vegetable Oils, and Surfactants on the Properties of Sodium-Alginate-Based Edible Coatings.
Effect of Presence and Concentration of Plasticizers, Vegetable Oils, and Surfactants on the Properties of Sodium-Alginate-Based Edible Coatings.
International journal of molecular sciences (2018-03-07)
Tugce Senturk Parreidt, Michael Schott, Markus Schmid, Kajetan Müller
ABSTRACT
Achieving high quality of a coated food product is mostly dependent on the characteristics of the food material to be coated, the properties of the components in the coating solution, and the obtained coating material. In the present study, usability and effectiveness of various components as well as their concentrations were assessed to produce an effective coating material. For this purpose, different concentrations of gelling agent (sodium alginate 0-3.5%, w/w), plasticizers (glycerol and sorbitol (0-20%, w/w), surfactants (tween 40, tween 80, span 60, span 80, lecithin (0-5%, w/w), and vegetable oils (sunflower oil, olive oil, rapeseed oil (0-5%, w/w) were used to prepare edible coating solutions. Formulations were built gradually, and characteristics of coatings were evaluated by analyzing surface tension values and its polar and dispersive components, emulsion droplet size, and optical appearance in microscopic scale. The results obtained showed that 1.25% sodium alginate, 2% glycerol, 0.2% sunflower oil, 1% span 80, and 0.2% tween 40 or tween 80 can be used in formulation to obtain an effective coating for hydrophobic food surfaces. Three formulations were designed, and their stability (emulsion droplet size, optical characteristics, and creaming index) and wettability tests on strawberry showed that they could be successfully used in coating applications.
Sigma-Aldrich
Sorbitan monostearate, meets FCC analytical specifications