MilliporeSigma
Activators and targets.
Nature (1990-07-26)
M Ptashne, A A Gann
ABSTRACT
Proteins that activate genes are quite disparate in character; in particular, some work 'universally' and others do not. A simple model can accommodate most of the recently published results.
Sigma-Aldrich
GAL4 [(1-147) - AH] from Saccharomyces cerevisiae, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)